COM448654332 Combi-Blocks, Inc.

Potassium 2-(oxiran-2-yl)ethyltrifluoroborate

  • MDL number MFCD11052752
  • Cas Number : 608140-72-3
  • Molecular Weight : 178.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0720

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0720
Availability details for
Quantity
  • Availability
  • Add to Cart