COM448654352 Combi-Blocks, Inc.

2-(3-(tert-Butoxycarbonylamino)-2-oxopyrrolidin-1-yl)acetic acid

  • MDL number MFCD09878668
  • Cas Number : 116339-45-8
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0740

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0740
Availability details for
Quantity
  • Availability
  • Add to Cart