COM448654395 Combi-Blocks, Inc.

Ethyl 2-cyano-2-(3,4-dimethoxyphenyl)acetate

  • MDL number MFCD03412574
  • Cas Number : 36848-69-8
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0784

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0784
Availability details for
Quantity
  • Availability
  • Add to Cart