COM448654431 Combi-Blocks, Inc.

4-(2-Methyl-1,3-thiazol-4-yl)benzoic acid

  • MDL number MFCD02225088
  • Cas Number : 294620-60-3
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0821
Availability details for
Quantity
  • Availability
  • Add to Cart