COM448654439 Combi-Blocks, Inc.

N-(4-Nitrobenzoyl)-beta-alanine

  • MDL number MFCD00009794
  • Cas Number : 59642-21-6
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0830
Availability details for
Quantity
  • Availability
  • Add to Cart