COM448654463 Combi-Blocks, Inc.

5,6,7,8-Tetrahydro-2,6-naphthyridin-1(2H)-one

  • MDL number MFCD12026312
  • Cas Number : 1170830-85-9
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0854
Availability details for
Quantity
  • Availability
  • Add to Cart