COM448654510 Combi-Blocks, Inc.

N-Me-dl-tyr-ome hcl

  • MDL number MFCD03788157
  • Molecular Weight : 245.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0902
Availability details for
Quantity
  • Availability
  • Add to Cart