COM448654746 Combi-Blocks, Inc.

3,4-Dimethoxy-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD14705135
  • Cas Number : 138490-96-7
  • Molecular Weight : 234.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1144

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1144
Availability details for
Quantity
  • Availability
  • Add to Cart