COM448654811 Combi-Blocks, Inc.

3-(Phenethylsulfonyl)pyrrolidine

  • MDL number MFCD14585301
  • Cas Number : 1206969-01-8
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1211

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1211
Availability details for
Quantity
  • Availability
  • Add to Cart