COM448654880 Combi-Blocks, Inc.

3-(5-Fluoropyridin-2-yl)acrylic acid hydrochloride

  • MDL number MFCD23135525
  • Cas Number : 917760-91-9
  • Molecular Weight : 203.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1281

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1281
Availability details for
Quantity
  • Availability
  • Add to Cart