COM448654927 Combi-Blocks, Inc.

4-Methoxy-1h-indazole-6-carboxylic acid

  • MDL number MFCD11845641
  • Cas Number : 1167055-67-5
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1328

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1328
Availability details for
Quantity
  • Availability
  • Add to Cart