COM448654951 Combi-Blocks, Inc.

1,2:5,6-Di-O-isopropylidene-alpha-D-gulofuranose

  • MDL number MFCD00069693
  • Cas Number : 14686-89-6
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1354
Availability details for
Quantity
  • Availability
  • Add to Cart