COM448655016 Combi-Blocks, Inc.

8-Boc-3,8-diazabicyclo[4.2.0]octane

  • MDL number MFCD09056795
  • Cas Number : 848591-80-0
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1423

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1423
Availability details for
Quantity
  • Availability
  • Add to Cart