COM448655055 Combi-Blocks, Inc.

2-(9-Oxoxanthen-2-yl)propionic acid

  • MDL number MFCD01741547
  • Cas Number : 30087-33-3
  • Molecular Weight : 268.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1463

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1463
Availability details for
Quantity
  • Availability
  • Add to Cart