COM448655063 Combi-Blocks, Inc.

2-Hydroxy-5-trifluoromethyl-nicotinonitrile

  • MDL number MFCD09909905
  • Cas Number : 124432-69-5
  • Molecular Weight : 188.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1471

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1471
Availability details for
Quantity
  • Availability
  • Add to Cart