COM448655066 Combi-Blocks, Inc.

4-Amino-2-methylthio-5-nitrothiazole

  • MDL number MFCD06658048
  • Cas Number : 127346-42-3
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1474

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1474
Availability details for
Quantity
  • Availability
  • Add to Cart