COM448655071 Combi-Blocks, Inc.

1-Bromo-2-(difluoromethyl)-4-methoxybenzene

  • MDL number MFCD14698634
  • Cas Number : 1214329-81-3
  • Molecular Weight : 237.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1480

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1480
Availability details for
Quantity
  • Availability
  • Add to Cart