COM448655131 Combi-Blocks, Inc.

Europium(iii) oxalate hydrate

  • MDL number MFCD00150143
  • Cas Number : 152864-32-9
  • Molecular Weight : 586.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1542
Availability details for
Quantity
  • Availability
  • Add to Cart