COM448655216 Combi-Blocks, Inc.

Benzyl pyrrolidin-3-ylcarbamate hydrochloride

  • MDL number MFCD11101361
  • Cas Number : 553672-38-1
  • Molecular Weight : 256.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1628

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1628
Availability details for
Quantity
  • Availability
  • Add to Cart