COM448655275 Combi-Blocks, Inc.

N-Methyl-n-[3-(2-methyl-1,3-thiazol-4-yl)benzyl]amine

  • MDL number MFCD07368505
  • Cas Number : 852180-43-9
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1687

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1687
Availability details for
Quantity
  • Availability
  • Add to Cart