COM448655308 Combi-Blocks, Inc.

[4-(Piperidin-1-ylmethyl)phenyl]methanol

  • MDL number MFCD07772863
  • Cas Number : 91271-62-4
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1721
Availability details for
Quantity
  • Availability
  • Add to Cart