COM448655391 Combi-Blocks, Inc.

1-Fluoro-1,1,3-trichloropropane

  • MDL number MFCD00082624
  • Cas Number : 818-99-5
  • Molecular Weight : 165.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1806
Availability details for
Quantity
  • Availability
  • Add to Cart