COM448655398 Combi-Blocks, Inc.

Tetraisopropyl orthosilicate

  • MDL number MFCD00026379
  • Cas Number : 1992-48-9
  • Molecular Weight : 264.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1814

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1814
Availability details for
Quantity
  • Availability
  • Add to Cart