COM448655402 Combi-Blocks, Inc.

5-Nitrofluorescein

  • MDL number MFCD00059693
  • Cas Number : 3326-35-0
  • Molecular Weight : 377.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 96
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1818
Availability details for
Quantity
  • Availability
  • Add to Cart