COM448655458 Combi-Blocks, Inc.

2-Chloro-5-trifluoromethyl-benzamidine

  • MDL number MFCD05662902
  • Cas Number : 885963-61-1
  • Molecular Weight : 222.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1876
Availability details for
Quantity
  • Availability
  • Add to Cart