COM448655533 Combi-Blocks, Inc.

1H,1H-Perfluoro(2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecan-1-ol)

  • MDL number MFCD08460480
  • Cas Number : 141977-66-4
  • Molecular Weight : 814.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1955
Availability details for
Quantity
  • Availability
  • Add to Cart