COM448655536 Combi-Blocks, Inc.

Ormetoprim

  • MDL number MFCD00057747
  • Cas Number : 6981-18-6
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1958
Availability details for
Quantity
  • Availability
  • Add to Cart