COM448655543 Combi-Blocks, Inc.

Vinyl n-octanoate

  • MDL number MFCD00048915
  • Cas Number : 818-44-0
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-1968
Availability details for
Quantity
  • Availability
  • Add to Cart