COM448655573 Combi-Blocks, Inc.

4-Methylisoquinolin-5-amine

  • MDL number MFCD12922847
  • Cas Number : 194032-18-3
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2002
Availability details for
Quantity
  • Availability
  • Add to Cart