COM448655583 Combi-Blocks, Inc.

Phenyl beta-d-glucopyranoside hydrate

  • MDL number MFCD03410292
  • Cas Number : 636565-38-3
  • Molecular Weight : 274.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2014

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2014
Availability details for
Quantity
  • Availability
  • Add to Cart