COM448655614 Combi-Blocks, Inc.

1-(2-Methylphenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD11037063
  • Cas Number : 151157-47-0
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2048
Availability details for
Quantity
  • Availability
  • Add to Cart