COM448655621 Combi-Blocks, Inc.

2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde

  • MDL number MFCD09702359
  • Cas Number : 879896-54-5
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2056

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2056
Availability details for
Quantity
  • Availability
  • Add to Cart