COM448655633 Combi-Blocks, Inc.

1,4-Dimethyladamantane

  • MDL number MFCD09743483
  • Cas Number : 16267-35-9
  • Molecular Weight : 164.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2068

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2068
Availability details for
Quantity
  • Availability
  • Add to Cart