COM448655661 Combi-Blocks, Inc.

Salicylic acid 4-tert-butylphenyl ester

  • MDL number MFCD00045759
  • Cas Number : 87-18-3
  • Molecular Weight : 270.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2097
Availability details for
Quantity
  • Availability
  • Add to Cart