COM448655709 Combi-Blocks, Inc.

2-(Fmoc-amino)-4-chlorobenzoic acid

  • MDL number MFCD02682157
  • Cas Number : 332121-92-3
  • Molecular Weight : 393.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2147
Availability details for
Quantity
  • Availability
  • Add to Cart