COM448655713 Combi-Blocks, Inc.

3-Fluoroquinolin-8-amine

  • MDL number MFCD18802884
  • Cas Number : 846038-33-3
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2152

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2152
Availability details for
Quantity
  • Availability
  • Add to Cart