COM448655734 Combi-Blocks, Inc.

Hs-dpeg(4)-co-otbu

  • MDL number MFCD11041133
  • Cas Number : 564476-33-1
  • Molecular Weight : 338.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2174

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2174
Availability details for
Quantity
  • Availability
  • Add to Cart