COM448655788 Combi-Blocks, Inc.

Guanosine 2'-3'-monophosphate disodium salt

  • MDL number MFCD00213394
  • Molecular Weight : 814.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2230
Availability details for
Quantity
  • Availability
  • Add to Cart