COM448655833 Combi-Blocks, Inc.

1-(Bromomethyl)-3-cyclopropylbenzene

  • MDL number MFCD15527185
  • Cas Number : 1260850-05-2
  • Molecular Weight : 211.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2276
Availability details for
Quantity
  • Availability
  • Add to Cart