COM448655842 Combi-Blocks, Inc.

5-Bromo-4-chloro-3-indolyl-n-acetyl-beta-d-galactosaminide

  • MDL number MFCD00083260
  • Cas Number : 129572-48-1
  • Molecular Weight : 449.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2286
Availability details for
Quantity
  • Availability
  • Add to Cart