COM448655882 Combi-Blocks, Inc.

N-Boc-l-(+)-penicillamine

  • MDL number MFCD02682558
  • Cas Number : 110763-40-1
  • Molecular Weight : 249.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2330

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2330
Availability details for
Quantity
  • Availability
  • Add to Cart