COM448655930 Combi-Blocks, Inc.

N-Benzyl-n-(2-chloroethyl)amine hydrochloride

  • MDL number MFCD00034901
  • Cas Number : 6288-63-7
  • Molecular Weight : 206.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2379

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2379
Availability details for
Quantity
  • Availability
  • Add to Cart