COM448656003 Combi-Blocks, Inc.

Potassium ((1-[(tert-butoxy)carbonyl]azetidin-3-yl)methyl)trifluoroboranuide

  • MDL number MFCD18712403
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2458

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2458
Availability details for
Quantity
  • Availability
  • Add to Cart