COM448656022 Combi-Blocks, Inc.

L-Inosine

  • MDL number MFCD00673221
  • Cas Number : 21138-24-9
  • Molecular Weight : 268.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2477

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2477
Availability details for
Quantity
  • Availability
  • Add to Cart