COM448656041 Combi-Blocks, Inc.

1,2:5,6-Di-o-isopropylidene-3-o-p-toluenesulfonyl-alpha-d-allofuranose

  • MDL number MFCD02094244
  • Cas Number : 13964-21-1
  • Molecular Weight : 412.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2498
Availability details for
Quantity
  • Availability
  • Add to Cart