COM448656064 Combi-Blocks, Inc.

3-Hydroxyquinolin-2-one

  • MDL number MFCD08705722
  • Cas Number : 26386-86-7
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2522
Availability details for
Quantity
  • Availability
  • Add to Cart