COM448656088 Combi-Blocks, Inc.

Meso-tetra(3-methylphenyl) porphine

  • MDL number MFCD02114464
  • Cas Number : 50849-45-1
  • Molecular Weight : 670.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2548

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2548
Availability details for
Quantity
  • Availability
  • Add to Cart