COM448656089 Combi-Blocks, Inc.

4-Phenyl-5-(trifluoromethyl)thiophene-2-carboxaldehyde

  • MDL number MFCD06808643
  • Cas Number : 874832-11-8
  • Molecular Weight : 256.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2549

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2549
Availability details for
Quantity
  • Availability
  • Add to Cart