COM448656107 Combi-Blocks, Inc.

Benzyl 1-benzyl-4-methylpiperazine-2-carboxylate

  • MDL number MFCD09037846
  • Cas Number : 1447605-99-3
  • Molecular Weight : 324.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2569
Availability details for
Quantity
  • Availability
  • Add to Cart