COM448656217 Combi-Blocks, Inc.

1-(4-Fluoro-phenyl)-2-pyridin-4-yl-ethanone

  • MDL number MFCD03069461
  • Cas Number : 6576-05-2
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-2689
Availability details for
Quantity
  • Availability
  • Add to Cart